2-Toluenethiol ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Thiocresol, 1-Mercapto-2-methylbenzene, o-Toluenethiol, 2-Methylbenzene-1-thiol, 2-Methylbenzenethiol, o-Thiocresol, 2-Methylthiophenol

T0288-25ML T0288-100ML
TCIAT0288-25MLEA 2100 SEK
TCIAT0288-25ML TCIAT0288-100ML
2-Toluenethiol ≥97.0%
2-Toluenethiol
Formel: C₇H₈S
MW: 124,21 g/mol
Kokpunkt: 194°C
Smältpunkt: 4,5 °C
Densitet: 1,041 g/cm³ (20 °C)
Flampunkt: 64°C
Storage Temperature: Omgivande
MDL Number: MFCD00004838
CAS nummer: 137-06-4
EINECS: 205-276-8
ADR: 6.1,III
Merck Index: 14,09325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=895bfaf9f799911de5a7ee0d0837050b80dafd72&vid=895bfaf9f799911de5a7ee0d0837050b80dafd72&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR