4-Methoxyphenyl isocyanate ≥98.0%
Leverantör: TCI
Fara
Synonyms: 1-Isocyanato-4-methoxybenzene
I0440-25G I0440-5G
TCIAI0440-25GEA 1825 SEK
TCIAI0440-25G TCIAI0440-5G
4-Methoxyphenyl isocyanate ≥98.0%
4-Methoxyphenyl isocyanate
Formel: C₈H₇NO₂
MW: 149.15 g/mol
Kokpunkt: 133°C
Densitet: 1.1559
Flampunkt: 98°C
MDL Number: MFCD00002026
CAS nummer: 5416-93-3
EINECS: 226-513-1
UN: 2206
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0416afbf5eb8bdbb22c7e398abfcd68c6f17eda6&vid=0416afbf5eb8bdbb22c7e398abfcd68c6f17eda6&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %