1-Iodo-4,4,4-trifluorobutane ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Iodo-1,1,1-trifluorobutane, 1,1,1-Trifluoro-4-iodobutane

T2529-5G T2529-25G
TCIAT2529-5GEA 1075 SEK
TCIAT2529-5G TCIAT2529-25G
1-Iodo-4,4,4-trifluorobutane ≥98.0%
1-Iodo-4,4,4-trifluorobutane
Formel: C₄H₆F₃I
MW: 237,99 g/mol
MDL Number: MFCD00041543
CAS nummer: 461-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45b62e9fc639426a6b343620d390348182592e0d&vid=45b62e9fc639426a6b343620d390348182592e0d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR