Bifenyl-4-ol ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Hydroxibifenyl

P0201-25G P0201-500G P0201-100G
TCIAP0201-25GEA 230 SEK
TCIAP0201-25G TCIAP0201-500G TCIAP0201-100G
Bifenyl-4-ol ≥99.0%
Bifenyl-4-ol
Formel: C₁₂H₁₀O
MW: 170,21 g/mol
Kokpunkt: 321 °C (1013 hPa)
Smältpunkt: 162 °C
Densitet: 1,29 g/cm³ (20 °C)
MDL Number: MFCD00002347
CAS nummer: 92-69-3
EINECS: 202-179-2
UN: 3077
ADR: 9,III
Merck Index: 14,07305
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e71add6227be85a89082d92d7caaba5d2c0d47c&vid=7e71add6227be85a89082d92d7caaba5d2c0d47c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 164.0 to 168.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR