2-Fluorophenyl isocyanate ≥97.0%
Leverantör: TCI
|
Fara
|
I0338-25G
I0338-5G
TCIAI0338-25GEA
3000
SEK
TCIAI0338-25G
TCIAI0338-5G
2-Fluorophenyl isocyanate ≥97.0%
2-Fluorophenyl isocyanate
Formel:
C₇H₄FNO Kokpunkt: 65°C Densitet: 1,222 g/cm³ (20 °C) Flampunkt: 50°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00001996 CAS nummer: 16744-98-2 EINECS: 240-804-0 UN: 2478 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...