4'-Nitroacetophenone ≥98.0%
Leverantör: TCI
Varning
Synonyms: p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
N0111-25G N0111-500G N0111-100G
TCIAN0111-25GEA 215 SEK
TCIAN0111-25G TCIAN0111-500G TCIAN0111-100G
4'-Nitroacetophenone ≥98.0%
4'-Nitroacetophenone
Formel: C₈H₇NO₃
MW: 165.15 g/mol
Kokpunkt: 167°C
Flampunkt: 149
Storage Temperature: Omgivande
MDL Number: MFCD00007355
CAS nummer: 100-19-6
EINECS: 202-827-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f011bc6e5a711b6699e4d4e779efb6566a89760b&vid=f011bc6e5a711b6699e4d4e779efb6566a89760b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Solubility in Methanol very faint turbidity