4'-Fluoroacetofenon ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-(4-Fluorophenyl)ethanone, p-Fluoroacetophenone
F0261-25G
F0261-500G
TCIAF0261-25GEA
310
SEK
TCIAF0261-25G
TCIAF0261-500G
4'-Fluoroacetofenon ≥98.0%
4'-Fluoroacetofenon
Formel:
C₈H₇FO MW: 138,14 g/mol Kokpunkt: 77°C Densitet: 1,138 g/cm³ (20 °C) Flampunkt: 71°C Storage Temperature: Omgivande |
MDL Number:
MFCD00000354 CAS nummer: 403-42-9 EINECS: 206-960-9 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...