2'-Fluoroacetophenone ≥97.0%
Leverantör: TCI
Varning
Synonyms: 1-(2-Fluorophenyl)ethanone, o-Fluoroacetophenone
F0364-25G F0364-5G
TCIAF0364-25GEA 965 SEK
TCIAF0364-25G TCIAF0364-5G
2'-Fluoroacetophenone ≥97.0%
2'-Fluoroacetophenone
Formel: C₈H₇FO
Kokpunkt: 85°C
Densitet: 1,12 g/cm³ (20 °C)
Flampunkt: 61°C
Storage Temperature: Omgivande
MDL Number: MFCD00000320
CAS nummer: 445-27-2
EINECS: 207-156-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c7312bd377c6fef0107ddc0c3841623a03a2413&vid=2c7312bd377c6fef0107ddc0c3841623a03a2413&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %