1-Chlorotridecane ≥98.0%

Leverantör: TCI EUROPE
Varning

C2213-25G C2213-5G
TCIAC2213-25GEA 8700 SEK
TCIAC2213-25G TCIAC2213-5G
1-Chlorotridecane ≥98.0%
1-Chlorotridecane
Formel: C₁₃H₂₇Cl
MW: 218,81 g/mol
Kokpunkt: 126°C
CAS nummer: 822-13-9
EINECS: 212-488-4
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72eee1d3f36febc92bfe0775a5cc9d0e448cb68b&vid=72eee1d3f36febc92bfe0775a5cc9d0e448cb68b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR