(2-Methoxyethoxy)methyl chloride ≥95.0%
Leverantör: TCI
|
Fara
|
Synonyms:
1-Chloromethoxy-2-methoxyethane, 1-Chloro-2,5-dioxahexane, MEM chloride
M0680-25ML
M0680-500ML
TCIAM0680-25MLEA
1025
SEK
TCIAM0680-25ML
TCIAM0680-500ML
(2-Methoxyethoxy)methyl chloride ≥95.0%
(2-Methoxyethoxy)methyl chloride
Formel:
ClCH₂OCH₂CH₂OCH₃ Kokpunkt: 50 °C (1013 hPa) Densitet: 1,094 g/cm³ (20 °C) Flampunkt: 54°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00000888 CAS nummer: 3970-21-6 EINECS: 223-589-8 UN: 2929 ADR: 6.1,II |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Bis chloromethyl ether (GC) | max. 1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...