4'-Chloroacetophenone ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: p-Chloroacetophenone

C0033-25G C0033-500G C0033-100G
TCIAC0033-25GEA 215 SEK
TCIAC0033-25G TCIAC0033-500G TCIAC0033-100G
4'-Chloroacetophenone ≥97.0%
4'-Chloroacetophenone
Formel: C₈H₇ClO
MW: 154,6 g/mol
Kokpunkt: 232 °C (1013 hPa)
Smältpunkt: 18…20 °C
Densitet: 1,1952 g/cm³ (20 °C)
Flampunkt: 118°C
Storage Temperature: Omgivande
MDL Number: MFCD00000624
CAS nummer: 99-91-2
EINECS: 202-800-7
UN: 3416
ADR: 6.1,II
Merck Index: 14,02116
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75cd887c3e17f0f1ba1773eec2c6680261439357&vid=75cd887c3e17f0f1ba1773eec2c6680261439357&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR