4-Chlorobenzophenone ≥98.0%
Leverantör: TCI EUROPE
Synonyms: p-Chlorobenzophenone
C0136-25G C0136-500G
TCIAC0136-25GEA 215 SEK
TCIAC0136-25G TCIAC0136-500G
4-Chlorobenzophenone ≥98.0%
4-Chlorobenzophenone
Formel: C₁₃H₉ClO
MW: 216,67 g/mol
Kokpunkt: 170°C
Smältpunkt: 68…70 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000622
CAS nummer: 134-85-0
EINECS: 205-160-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99d6f8e346378d2259924a40ae216e48d649a3b1&vid=99d6f8e346378d2259924a40ae216e48d649a3b1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 78.0 deg-C
Solubility in hot Methanol almost transparency