1-Chloro-3,5-dibromobenzene ≥96.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 3,5-Dibromochlorobenzene, 1,3-Dibromo-5-chlorobenzene
D3465-25G D3465-5G
TCIAD3465-25GEA 1250 SEK
TCIAD3465-25G TCIAD3465-5G
1-Chloro-3,5-dibromobenzene ≥96.0%
1-Chloro-3,5-dibromobenzene
Formel: C₆H₃Br₂Cl
MW: 270,35 g/mol
Kokpunkt: 256°C
MDL Number: MFCD00070765
CAS nummer: 14862-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e41119f079c83d43e7ddc27b1276e30b12b34c1&vid=8e41119f079c83d43e7ddc27b1276e30b12b34c1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 96.0 %
Melting point 94.0 to 98.0 deg-C