2-Chloro-5-nitrophenol ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
C2077-25G
C2077-5G
TCIAC2077-25GEA
2675
SEK
TCIAC2077-25G
TCIAC2077-5G
2-Chloro-5-nitrophenol ≥98.0% (by GC, titration analysis)
2-Chloro-5-nitrophenol
Formel:
C₆H₄ClNO₃ MW: 173,56 g/mol Smältpunkt: 112°C |
MDL Number:
MFCD01571825 CAS nummer: 619-10-3 EINECS: 210-579-3 |
Specifikation testresultat
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 117.0 to 123.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...