2'-Chloroacetophenone ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1-(2-Chlorophenyl)ethanone, o-Chloroacetophenone
C1084-25G C1084-250G
TCIAC1084-25GEA 335 SEK
TCIAC1084-25G TCIAC1084-250G
2'-Chloroacetophenone ≥97.0%
2'-Chloroacetophenone
Formel: C₈H₇ClO
MW: 154,6 g/mol
Kokpunkt: 229°C
Densitet: 1,189 g/cm³ (20 °C)
Flampunkt: 92°C
Storage Temperature: Omgivande
MDL Number: MFCD00000560
CAS nummer: 2142-68-9
EINECS: 218-397-6
UN: 3416
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5e78d611a84ef16afb0b22fed9ce371e59d6423&vid=f5e78d611a84ef16afb0b22fed9ce371e59d6423&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %