4-Bromophenyl isocyanate ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 1-Bromo-4-isocyanatobenzene
B0636-25G B0636-5G
TCIAB0636-25GEA 5025 SEK
TCIAB0636-25G TCIAB0636-5G
4-Bromophenyl isocyanate ≥98.0%
4-Bromophenyl isocyanate
Formel: C₇H₄BrNO
MW: 198,02 g/mol
Kokpunkt: 226 °C (1013 hPa)
Smältpunkt: 42 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00002022
CAS nummer: 2493-02-9
EINECS: 219-662-9
UN: 2206
ADR: 6.1,III
Merck Index: 14,01430
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 39.0 to 43.0 deg-C