5-Bromo-α,α,α,2-tetrafluorotoluene ≥98.0%
Leverantör: TCI
Varning
Synonyms: 5-Bromo-2-fluorobenzotrifluoride, 1-Bromo-4-fluoro-3-(trifluoromethyl)benzene
B2872-25G B2872-5G
TCIAB2872-25GEA 1375 SEK
TCIAB2872-25G TCIAB2872-5G
5-Bromo-α,α,α,2-tetrafluorotoluene ≥98.0%
5-Bromo-α,α,α,2-tetrafluorotoluene
Formel: C₇H₃BrF₄
MW: 243 g/mol
Kokpunkt: 147°C
MDL Number: MFCD00042493
CAS nummer: 393-37-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e357b82ff5dfc4a3ba88640668d034c80fe64195&vid=e357b82ff5dfc4a3ba88640668d034c80fe64195&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %