4'-Bromoacetophenone ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: p-Bromoacetophenone, 1-(4-Bromophenyl)ethanone

B0537-25G B0537-500G
TCIAB0537-25GEA 215 SEK
TCIAB0537-25G TCIAB0537-500G
4'-Bromoacetophenone ≥98.0%
4'-Bromoacetophenone
Formel: C₈H₇BrO
MW: 199,05 g/mol
Kokpunkt: 255 °C (1013 hPa)
Smältpunkt: 49…52 °C
Densitet: 1,736 g/cm³ (20 °C)
Flampunkt: ≥ 100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000105
CAS nummer: 99-90-1
EINECS: 202-799-3
ADR: 6.1,II
Merck Index: 14,01403
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0762177456bb15031cf44947afe9798d7286cfe3&vid=0762177456bb15031cf44947afe9798d7286cfe3&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR