2-Bromo-5-chloroanisole ≥98.0%

Leverantör: TCI

Synonyms: 1-Bromo-4-chloro-2-methoxybenzene

B3646-25G B3646-5G
TCIAB3646-25GEA 1375 SEK
TCIAB3646-25G TCIAB3646-5G
2-Bromo-5-chloroanisole ≥98.0%
2-Bromo-5-chloroanisole
Formel: C₇H₆BrClO
MDL Number: MFCD03790889
CAS nummer: 174913-09-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17a45a64eff877bb86b2449a4b83deea4bc380a6&vid=17a45a64eff877bb86b2449a4b83deea4bc380a6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR