5-Bromo-m-xylene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Bromo-3,5-dimethylbenzene

B1792-25G B1792-5G
TCIAB1792-25GEA 590 SEK
TCIAB1792-25G TCIAB1792-5G
5-Bromo-m-xylene ≥98.0%
5-Bromo-m-xylene
Formel: C₈H₉Br
MW: 185,06 g/mol
Kokpunkt: 205°C
Densitet: 1,36 g/cm³ (20 °C)
Flampunkt: 87°C
Storage Temperature: Omgivande
MDL Number: MFCD00000087
CAS nummer: 556-96-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105e4a6fcec6564b4357f25c74e2a191d13b3c7b&vid=105e4a6fcec6564b4357f25c74e2a191d13b3c7b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR