4-Bromo-3-fluoroanisole ≥98.0%
Leverantör: TCI EUROPE
Varning
B3743-25G B3743-5G
TCIAB3743-25GEA 2800 SEK
TCIAB3743-25G TCIAB3743-5G
4-Bromo-3-fluoroanisole ≥98.0%
4-Bromo-3-fluoroanisole
Formel: C₇H₆BrFO
MW: 205,03 g/mol
Kokpunkt: 216°C
MDL Number: MFCD01310983
CAS nummer: 458-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63e4e4486890d344ae96f56e0f4ef47132bc077f&vid=63e4e4486890d344ae96f56e0f4ef47132bc077f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %