4-Bromo-α,α,α,3-tetrafluorotoluene ≥98.0%
Leverantör: TCI
Varning
B3783-25G B3783-5G
TCIAB3783-25GEA 2275 SEK
TCIAB3783-25G TCIAB3783-5G
4-Bromo-α,α,α,3-tetrafluorotoluene ≥98.0%
4-Bromo-α,α,α,3-tetrafluorotoluene
Formel: C₇H₃BrF₄
MW: 243 g/mol
Kokpunkt: 145°C
Storage Temperature: Kylskåp
MDL Number: MFCD00042497
CAS nummer: 40161-54-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d858b16e0bfb5797feba8836d9e9ad988a81b46&vid=3d858b16e0bfb5797feba8836d9e9ad988a81b46&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %