5-Bromo-o-toluidine ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 5-Bromo-2-methylaniline, 5-Bromo-o-toluidine, 2-Amino-4-bromotoluene
B3687-25G B3687-5G
TCIAB3687-25GEA 1475 SEK
TCIAB3687-25G TCIAB3687-5G
5-Bromo-o-toluidine ≥97.0% (by GC, titration analysis)
5-Bromo-o-toluidine
Formel: C₇H₈BrN
MW: 186,05 g/mol
Kokpunkt: 139°C
Smältpunkt: 33 °C
Densitet: 1,49 g/cm³ (25 °C)
MDL Number: MFCD00800678
CAS nummer: 39478-78-9
EINECS: 254-467-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14e986fd6ef2267efb51b5a6e336a82a0879100f&vid=14e986fd6ef2267efb51b5a6e336a82a0879100f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Green powder to lump
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %