4-Nitro-2-(trifluoromethyl)aniline ≥98.0%
Leverantör: TCI
Varning
A0805-1G A0805-25G A0805-5G
TCIAA0805-1GEA 215 SEK
TCIAA0805-1G TCIAA0805-25G TCIAA0805-5G
4-Nitro-2-(trifluoromethyl)aniline ≥98.0%
4-Nitro-2-(trifluoromethyl)aniline
Formel: C₇H₅F₃N₂O₂
Smältpunkt: 94…95 °C
Densitet: 1,754 g/cm³ (-123.15 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007365
CAS nummer: 121-01-7
EINECS: 204-443-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a5e0b90fd1cd71993556ba07b7a7b097bc1967f&vid=0a5e0b90fd1cd71993556ba07b7a7b097bc1967f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 94.0 deg-C