4-Aminofenol ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
A0384-25G A0384-500G
TCIAA0384-25GEA 215 SEK
TCIAA0384-25G TCIAA0384-500G
4-Aminofenol ≥98.0% (by GC, titration analysis)
4-Aminofenol
Formel: H₂NC₆H₄OH
Kokpunkt: 284 °C (1013 hPa)
Smältpunkt: ∼186 °C
Densitet: 1,3 g/cm³ (20 °C)
Flampunkt: (closed cup)
Storage Temperature: Kylskåp
MDL Number: MFCD00007869
CAS nummer: 123-30-8
EINECS: 204-616-2
UN: 2512
ADR: 6.1,III
Merck Index: 14,00462
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8525646da658b17e890315cb1875669562bfe8c9&vid=8525646da658b17e890315cb1875669562bfe8c9&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot HCl (1+3) almost transparency