4-Fluoro-2-(trifluoromethyl)aniline ≥97.0%

Leverantör: TCI
Varning

A1158-25G A1158-5G
TCIAA1158-25GEA 2025 SEK
TCIAA1158-25G TCIAA1158-5G
4-Fluoro-2-(trifluoromethyl)aniline ≥97.0%
4-Fluoro-2-(trifluoromethyl)aniline
Formel: C₇H₅F₄N
Kokpunkt: 90°C
Densitet: 1,38 g/cm³ (20 °C)
Flampunkt: 72°C
Storage Temperature: Kylskåp
MDL Number: MFCD00007831
CAS nummer: 393-39-5
EINECS: 206-886-7
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01afc8ba9d70d8b47261c9a01b76262c56bc31f5&vid=01afc8ba9d70d8b47261c9a01b76262c56bc31f5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR