4-Bromo-m-toluidine ≥98.0%
Leverantör: TCI
Varning
B1990-25G B1990-5G
TCIAB1990-25GEA 655 SEK
TCIAB1990-25G TCIAB1990-5G
4-Bromo-m-toluidine ≥98.0%
4-Bromo-m-toluidine
Formel: C₇H₈BrN
Kokpunkt: 240°C
MDL Number: MFCD00007828
CAS nummer: 6933-10-4
EINECS: 230-056-3
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd5d3bfc9057bcdb730836511d0face05853ce3&vid=dfd5d3bfc9057bcdb730836511d0face05853ce3&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Gray to Brown powder to crystaline
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C