4-Aminobenzophenone ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(4-Aminophenyl)(phenyl)methanone, p-Aminobenzophenone
A1140-25G
A1140-5G
TCIAA1140-25GEA
975
SEK
TCIAA1140-25G
TCIAA1140-5G
4-Aminobenzophenone ≥98.0% (by GC, titration analysis)
4-Aminobenzophenone
Formel:
C₁₃H₁₁NO MW: 197,24 g/mol Kokpunkt: 246 °C (1013 hPa) Smältpunkt: 118…121 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00007895 CAS nummer: 1137-41-3 EINECS: 214-506-6 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...