5-Fluoro-2-nitroaniline ≥98.0%
Leverantör: TCI EUROPE
Varning
F0789-25G F0789-5G
TCIAF0789-25GEA 2725 SEK
TCIAF0789-25G TCIAF0789-5G
5-Fluoro-2-nitroaniline ≥98.0%
5-Fluoro-2-nitroaniline
Formel: C₆H₅FN₂O₂
MW: 156,12 g/mol
MDL Number: MFCD00034065
CAS nummer: 2369-11-1
EINECS: 219-128-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40c0758a05a844a75a9b6109f42a7d7ea004c117&vid=40c0758a05a844a75a9b6109f42a7d7ea004c117&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C