α,α,α-Trifluoro-m-toluidine ≥99.0%
Leverantör: TCI
Fara
Synonyms: 3-(Trifluoromethyl)aniline, 3-Aminobenzotrifluoride
A0434-25G A0434-500G A0434-100G
TCIAA0434-25GEA 215 SEK
TCIAA0434-25G TCIAA0434-500G TCIAA0434-100G
α,α,α-Trifluoro-m-toluidine ≥99.0%
α,α,α-Trifluoro-m-toluidine
Formel: C₇H₆F₃N
MW: 161.13 g/mol
Kokpunkt: 187
Smältpunkt: 5
Flampunkt: 85°C
Storage Temperature: Omgivande
MDL Number: MFCD00007797
CAS nummer: 98-16-8
EINECS: 202-643-4
UN: 2948
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52faea201a124bd5662fa5eae0d2eba78e1e5266&vid=52faea201a124bd5662fa5eae0d2eba78e1e5266&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %