Dipropylamin ≥99.0%

Leverantör: TCI
Fara

Synonyms: N-Propyl-1-propanamine

D0930-25ML D0930-500ML D0930-100ML
TCIAD0930-25MLEA 215 SEK
TCIAD0930-25ML TCIAD0930-500ML TCIAD0930-100ML
Dipropylamin ≥99.0%
Dipropylamin
Formel: C₆H₁₅N
Kokpunkt: 108°C
Smältpunkt: -63°C
Densitet: 0,738 g/cm³ (20 °C)
Flampunkt: 7°C
Storage Temperature: Omgivande
MDL Number: MFCD00009362
CAS nummer: 142-84-7
EINECS: 205-565-9
UN: 2383
ADR: 3,II
Merck Index: 14,03343
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe2ba88a5aee0e8ff1b29cd20c759b3f6e5fe7fa&vid=fe2ba88a5aee0e8ff1b29cd20c759b3f6e5fe7fa&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR