N-Metyl-p-toluidin ≥97.0%
Leverantör: TCI
|
Fara
|
Synonyms:
N,4-Dimetylbenzenamin
M1315-25ML
M1315-250ML
TCIAM1315-25MLEA
685
SEK
TCIAM1315-25ML
TCIAM1315-250ML
N-Metyl-p-toluidin ≥97.0%
N-Metyl-p-toluidin
Formel:
C₈H₁₁N MW: 121.18 g/mol Kokpunkt: 212°C Flampunkt: 83°C |
MDL Number:
MFCD00025628 CAS nummer: 623-08-5 EINECS: 210-769-6 UN: 2810 ADR: 6.1 III |
|
Specifikation testresultat
Appearance | Colorless to Orange to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...