4-Nitrobenzyl alcohol ≥98.0%
Leverantör: TCI
Varning
Synonyms: (4-Nitrophenyl)methanol
N0180-25G N0180-100G
TCIAN0180-25GEA 215 SEK
TCIAN0180-25G TCIAN0180-100G
4-Nitrobenzyl alcohol ≥98.0%
4-Nitrobenzyl alcohol
Formel: C₇H₇NO₃
MW: 153.14 g/mol
Kokpunkt: 185°C
Storage Temperature: Omgivande
MDL Number: MFCD00007376
CAS nummer: 619-73-8
EINECS: 210-611-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5de2c9c03b32f2802a2b71a89eb44671d42e365f&vid=5de2c9c03b32f2802a2b71a89eb44671d42e365f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C