(2,3,5,6-Tetrafluorofenyl)metanol ≥96.0%

Leverantör: TCI EUROPE
Varning

Synonyms: (2,3,5,6-Tetrafluorophenyl)methanol

T1884-1G T1884-5G
TCIAT1884-1GEA 325 SEK
TCIAT1884-1G TCIAT1884-5G
(2,3,5,6-Tetrafluorofenyl)metanol ≥96.0%
(2,3,5,6-Tetrafluorofenyl)metanol
Formel: C₇H₄F₄O
MW: 180,1 g/mol
MDL Number: MFCD00792428
CAS nummer: 4084-38-2
EINECS: 443-840-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87403080fcefe9e6849b279fdbdc45b000abd9f1&vid=87403080fcefe9e6849b279fdbdc45b000abd9f1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR