α-Bromo-3-(trifluoromethoxy)toluene ≥97.0%
Leverantör: TCI
Fara
Synonyms: m-(Trifluoromethoxy)benzyl bromide, 1-(Bromomethyl)-3-(trifluoromethoxy)benzene, 3-(Trifluoromethoxy)benzyl bromide
T2730-1G T2730-5G
TCIAT2730-1GEA 415 SEK
TCIAT2730-1G TCIAT2730-5G
α-Bromo-3-(trifluoromethoxy)toluene ≥97.0%
α-Bromo-3-(trifluoromethoxy)toluene
Formel: C₈H₆BrF₃O
MW: 255.03 g/mol
Densitet: 1.572
Flampunkt: 89°C
MDL Number: MFCD00061271
CAS nummer: 159689-88-0
UN: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b1b54f196cb9cc2b72d72184b37b9550e57574a&vid=0b1b54f196cb9cc2b72d72184b37b9550e57574a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %