3,5-Difluorobenzyl bromide ≥98.0%

Leverantör: TCI
Fara

Synonyms: α-Bromo-3,5-difluorotoluene

D2631-25G D2631-5G
TCIAD2631-25GEA 2450 SEK
TCIAD2631-25G TCIAD2631-5G
3,5-Difluorobenzyl bromide ≥98.0%
3,5-Difluorobenzyl bromide
Formel: C₇H₅BrF₂
Kokpunkt: 65°C
Densitet: 1,6 g/cm³ (20 °C)
Flampunkt: 81°C
MDL Number: MFCD00010304
CAS nummer: 141776-91-2
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=913dfd07500f4ea131557cf72d2527f0b07a91ab&vid=913dfd07500f4ea131557cf72d2527f0b07a91ab&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR