2,6-Difluorobenzylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
D1847-25G D1847-5G
TCIAD1847-25GEA 2700 SEK
TCIAD1847-25G TCIAD1847-5G
2,6-Difluorobenzylamine ≥98.0% (by GC, titration analysis)
2,6-Difluorobenzylamine
Formel: C₇H₇F₂N
MW: 143,14 g/mol
Kokpunkt: 177°C
Densitet: 1,19 g/cm³ (20 °C)
Flampunkt: 65°C
Storage Temperature: Omgivande
MDL Number: MFCD00010144
CAS nummer: 69385-30-4
EINECS: 273-983-9
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c083cdad8709ba4e9bc083e71e74263036f9dca&vid=0c083cdad8709ba4e9bc083e71e74263036f9dca&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %