Azelaic acid ≥80.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid
A0561-25G A0561-500G
TCIAA0561-25GEA 215 SEK
TCIAA0561-25G TCIAA0561-500G
Azelaic acid ≥80.0%
Azelaic acid
Formel: C₉H₁₆O₄
MW: 188,22 g/mol
Kokpunkt: 360°C
Smältpunkt: 106 °C
Densitet: 1,0287 g/cm³ (20 °C)
Flampunkt: 215 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004432
CAS nummer: 123-99-9
EINECS: 204-669-1
Merck Index: 14,00905
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cc197c25358a606244fcd69e5626dfb080e269d&vid=2cc197c25358a606244fcd69e5626dfb080e269d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 80.0 %
Melting point 102.0 to 105.0 deg-C