1,9-Decadiene ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Deca-1,9-diene

D0245-25ML D0245-5ML
TCIAD0245-25MLEA 1500 SEK
TCIAD0245-25ML TCIAD0245-5ML
1,9-Decadiene ≥98.0%
1,9-Decadiene
Formel: C₁₀H₁₈
MW: 138,25 g/mol
Kokpunkt: 165°C
Smältpunkt: –71,8 °C
Densitet: 0,7602 g/cm³ (20 °C)
Flampunkt: 41°C
Storage Temperature: Omgivande
MDL Number: MFCD00008670
CAS nummer: 1647-16-1
EINECS: 216-711-6
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91e47a4af13a13da2a9258d451a62a91d1f68572&vid=91e47a4af13a13da2a9258d451a62a91d1f68572&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR