1,6-Heptadiene ≥99.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Hepta-1,6-diene

H0564-1ML H0564-5ML
TCIAH0564-1MLEA 1025 SEK
TCIAH0564-1ML TCIAH0564-5ML
1,6-Heptadiene ≥99.0%
1,6-Heptadiene
Formel: C₇H₁₂
MW: 96,17 g/mol
Kokpunkt: 90°C
Smältpunkt: –129 °C
Densitet: 0,7142 g/cm³ (20 °C)
Flampunkt: –10°C
MDL Number: MFCD00008667
CAS nummer: 3070-53-9
UN: 3295
ADR: 3,II

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR