Hexametylen-1,6-diisocyanat ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Hexamethylene diisocyanate

H0324-25ML H0324-500ML H0324-100ML
TCIAH0324-25MLEA 255 SEK
TCIAH0324-25ML TCIAH0324-500ML TCIAH0324-100ML
Hexametylen-1,6-diisocyanat ≥98.0%
Hexametylen-1,6-diisocyanat
Formel: C₈H₁₂N₂O₂
MW: 168,2 g/mol
Kokpunkt: 255°C
Smältpunkt: –67 °C
Densitet: 1,05 g/cm³ (20 °C)
Flampunkt: 138°C
Storage Temperature: Kylskåp
MDL Number: MFCD00002047
CAS nummer: 822-06-0
EINECS: 212-485-8
UN: 2281
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f64a050b16dcbfe2c9afb36e0e45269e6d2e6f9b&vid=f64a050b16dcbfe2c9afb36e0e45269e6d2e6f9b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR