1,4-Pentadiene ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Penta-1,4-diene

P0038-10ML P0038-25ML
TCIAP0038-10MLEA 1800 SEK
TCIAP0038-10ML TCIAP0038-25ML
1,4-Pentadiene ≥98.0%
1,4-Pentadiene
Formel: C₅H₈
MW: 68,12 g/mol
Kokpunkt: 26°C
Smältpunkt: –148,3 °C
Densitet: 0,6608 g/cm³ (20 °C)
Flampunkt: –20°C
Storage Temperature: Frys
MDL Number: MFCD00008665
CAS nummer: 591-93-5
EINECS: 209-736-9
UN: 3295
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c302cfd43d2d36e25fe4810f6dccbff632a0c82&vid=8c302cfd43d2d36e25fe4810f6dccbff632a0c82&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR