p-Terphenyl ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Phenyl-1,1'-biphenyl, 1,4-Diphenylbenzene, 4-Phenylbiphenyl, 1,4-Terphenyl
T0020-25G
T0020-500G
T0020-100G
TCIAT0020-25GEA
375
SEK
TCIAT0020-25G
TCIAT0020-500G
TCIAT0020-100G
p-Terphenyl ≥99.0%
p-Terphenyl
Formel:
C₁₈H₁₄ MW: 230,31 g/mol Kokpunkt: 389°C Smältpunkt: 210…211 °C Densitet: 1,234 g/cm³ (0 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00003061 CAS nummer: 92-94-4 EINECS: 202-205-2 ADR: 9,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 211.0 to 216.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...