2-Metylhydrokinon ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Methylhydroquinone

M0342-25G M0342-500G M0342-100G
TCIAM0342-25GEA 285 SEK
TCIAM0342-25G TCIAM0342-500G TCIAM0342-100G
2-Metylhydrokinon ≥98.0%
2-Metylhydrokinon
Formel: C₇H₈O₂
MW: 124,14 g/mol
Kokpunkt: 285 °C (1013 hPa)
Smältpunkt: 123…126 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002345
CAS nummer: 95-71-6
EINECS: 202-443-7
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f94420ed7877288a6dcbfec8ff27d4bc946693ea&vid=f94420ed7877288a6dcbfec8ff27d4bc946693ea&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Redox method) min. 98.0 %
Melting point 126.0 to 130.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR