2',5'-Dihydroxyacetophenone ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-(2,5-Dihydroxyphenyl)ethan-1-one, 1-(2,5-Dihydroxyphenyl)ethanone, 2,5-Dihydroxyacetophenone

D1955-25G D1955-5G
TCIAD1955-25GEA 2225 SEK
TCIAD1955-25G TCIAD1955-5G
2',5'-Dihydroxyacetophenone ≥98.0%
2',5'-Dihydroxyacetophenone
Formel: C₈H₈O₃
MW: 152,15 g/mol
Kokpunkt: 296 °C (1013 hPa)
Smältpunkt: 199…202 °C
MDL Number: MFCD00002343
CAS nummer: 490-78-8
EINECS: 207-716-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ced0cd31753b8f35f42549d2812b024ddf8e3b8&vid=1ced0cd31753b8f35f42549d2812b024ddf8e3b8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 203.0 to 207.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR