2,5-Difluorobenzyl alcohol ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
(2,5-Difluorophenyl)methanol
D2818-5G
TCIAD2818-5GEA
1225
SEK
TCIAD2818-5G
2,5-Difluorobenzyl alcohol ≥98.0%
2,5-Difluorobenzyl alcohol
Formel:
C₇H₆F₂O MW: 144.12 g/mol Kokpunkt: 197 Smältpunkt: 9 Flampunkt: 93°C |
MDL Number:
MFCD00010627 CAS nummer: 75853-20-2 EINECS: 278-325-4 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...