Terephthalonitrile ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,4-Benzenedicarbonitrile, Terephthalodinitrile, Benzene-1,4-dicarbonitrile, 1,4-Dicyanobenzene

T0016-25G T0016-500G T0016-100G
TCIAT0016-25GEA 285 SEK
TCIAT0016-25G TCIAT0016-500G TCIAT0016-100G
Terephthalonitrile ≥98.0%
Terephthalonitrile
Formel: C₈H₄N₂
MW: 128,13 g/mol
Smältpunkt: 223…226 °C
Densitet: 1,29 g/cm³ (20 °C)
MDL Number: MFCD00001810
CAS nummer: 623-26-7
EINECS: 210-783-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c581ac59a695b0ab6207c55a53413d9d6a52cff&vid=7c581ac59a695b0ab6207c55a53413d9d6a52cff&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (GC) min. 98.0 %
Melting point 223.0 to 230.0 deg-C
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR