2',5'-Dichloroacetophenone ≥98.0%

Leverantör: TCI EUROPE

D2502-25G D2502-5G
TCIAD2502-25GEA 605 SEK
TCIAD2502-25G TCIAD2502-5G
2',5'-Dichloroacetophenone ≥98.0%
2',5'-Dichloroacetophenone
Formel: C₈H₆Cl₂O
MW: 189,04 g/mol
Kokpunkt: 128°C
Smältpunkt: 14°C
Densitet: 1,352 g/cm³ (17.5 °C)
MDL Number: MFCD00000607
CAS nummer: 2476-37-1
EINECS: 219-605-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=448325df5a05fe980d6d1edfb68e3d74caed01fd&vid=448325df5a05fe980d6d1edfb68e3d74caed01fd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR