2,5-Dichloro-α,α,α-trifluorotoluene ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1,4-Dichloro-2-(trifluoromethyl)benzene, 2,5-Dichlorobenzotrifluoride

D1318-25G D1318-5G
TCIAD1318-25GEA 1675 SEK
TCIAD1318-25G TCIAD1318-5G
2,5-Dichloro-α,α,α-trifluorotoluene ≥98.0%
2,5-Dichloro-α,α,α-trifluorotoluene
Formel: C₇H₃Cl₂F₃
MW: 215 g/mol
Kokpunkt: 181 °C (1013 hPa)
Smältpunkt: 11 °C
Densitet: 1,5009 g/cm³ (20 °C)
Flampunkt: 76°C
MDL Number: MFCD00000608
CAS nummer: 320-50-3
EINECS: 206-276-0
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68b200ae974072665010801c277fa06e5a73cc54&vid=68b200ae974072665010801c277fa06e5a73cc54&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR