3',5'-Difluoroacetophenone ≥97.0%

Leverantör: TCI
Varning

D3425-25G D3425-5G
TCIAD3425-25GEA 1800 SEK
TCIAD3425-25G TCIAD3425-5G
3',5'-Difluoroacetophenone ≥97.0%
3',5'-Difluoroacetophenone
Formel: C₈H₆F₂O
MDL Number: MFCD00042489
CAS nummer: 123577-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b372a70cb05f5d312fd6de1a56b1bdb512d2c4ff&vid=b372a70cb05f5d312fd6de1a56b1bdb512d2c4ff&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR